Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sirt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127351.1 | NP_001120823.1 | 6 | 901 | ||
NM_001177804.1 | NP_001171275.1 | 6 | 893 | |||
NM_022433.2 | NP_071878.2 | 6 | 946 | |||
GenBank mRNA | AF299338.1 | - | 6 | 946 | ||
AF299339.1 | - | 6 | 901 | |||
AK075861.1 | - | 6 | 875 | |||
BC025878.1 | - | 6 | 845 | 5253854 | ||
BF681854.1 | - | 1 | 127 | |||
CT010402.1 | - | 5 | 652 | |||
EU886466.1 | - | 6 | 891 | |||
FJ621493.1 | - | 6 | 889 |