Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nisch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022656.2 | NP_073147.2 | 20 | 4309 | ||
GenBank mRNA | AF315344.1 | - | 16 | 3762 | ||
AK122412.1 | - | 18 | 3807 | |||
AK191277.1 | - | 1 | 112 | |||
AK195831.1 | - | 1 | 112 | |||
AK198821.1 | - | 1 | 112 | |||
AY032852.1 | - | 3 | 444 | |||
BC089329.1 | - | 15 | 3528 | |||
BC108364.1 | - | 15 | 3504 | |||
BC144980.1 | - | 16 | 3638 |