Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Svep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022814.2 | NP_073725.2 | 38 | 9508 | ||
GenBank mRNA | AF206329.1 | - | 38 | 9508 | ||
AK014693.1 | - | 1 | 392 | |||
AK136952.1 | - | 1 | 1778 | |||
BC008135.1 | - | 1 | 595 |