Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199118.1 | NP_001186047.1 | 20 | 2996 | ||
NM_022883.3 | NP_075021.1 | 20 | 3073 | |||
GenBank mRNA | AF286724.1 | - | 20 | 3010 | ||
AK033647.1 | - | 20 | 3024 | |||
AK049363.1 | - | 20 | 2989 | |||
AK163833.1 | - | 20 | 2988 | |||
AK220228.1 | - | 20 | 3073 |