Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pes1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022889.3 | NP_075027.1 | 6 | 629 | ||
GenBank mRNA | AF289539.1 | - | 6 | 589 | ||
AK029075.1 | - | 6 | 629 | |||
AK088831.1 | - | 6 | 607 | |||
BC004844.1 | - | 6 | 614 | 2649598 | ||
BC011142.1 | - | 6 | 612 | 4207164 |