Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfand6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022985.6 | NP_075361.2 | 4, 5 | 490 | ||
GenBank mRNA | AJ251508.1 | - | 396 | |||
AK002775.1 | - | 4, 5 | 491 | |||
AK009403.1 | - | 3, 4 | 348 | |||
AK144902.1 | - | 3, 4 | 340 | |||
BC010683.1 | - | 3, 4 | 351 | 2647905 |