Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irak1bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168240.1 | NP_001161712.1 | 1 | 476 | ||
NM_022986.4 | NP_075362.3 | 1 | 476 | |||
GenBank mRNA | AF093135.1 | - | 1 | 259 | ||
AJ242722.1 | - | 1 | 101 | |||
AK013036.2 | - | 1 | 281 | |||
AK014712.1 | - | 1 | 253 | |||
AK020170.1 | - | 1 | 479 | |||
AK020448.1 | - | 1 | 251 | |||
AK045436.1 | - | 1 | 272 | |||
AK076594.1 | - | 1 | 284 | |||
BC046301.1 | - | 1 | 270 | |||
BC061099.1 | - | 1 | 263 | 6742251 | ||
BY719456.1 | - | 1 | 476 |