Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Recql | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204906.1 | NP_001191835.1 | 3 | 236 | ||
NM_001204907.1 | NP_001191836.1 | 3 | 236 | |||
NM_023042.3 | NP_075529.2 | 3 | 236 | |||
GenBank mRNA | AB017104.1 | - | 2 | 197 | ||
AB017105.1 | - | 2 | 189 | |||
AK137589.1 | - | 3 | 236 | |||
AK138325.1 | - | 3 | 192 | |||
AK138721.1 | - | 3 | 229 | |||
AK148430.1 | - | 3 | 224 | |||
AK164344.1 | - | 3 | 206 | |||
BC014735.1 | - | 2 | 168 | 4459244 |