Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpo7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023045.2 | NP_075532.1 | 4 | 482 | ||
GenBank mRNA | AJ297360.1 | - | 4 | 388 | ||
AK028303.1 | - | 2 | 227 | |||
AK035775.1 | - | 4 | 407 | |||
AK036837.1 | - | 2 | 227 | |||
AK041768.1 | - | 4 | 390 | |||
AK051105.1 | - | 4 | 404 | |||
AK122361.1 | - | 3 | 367 | |||
AK143729.1 | - | 4 | 451 | |||
AK164595.1 | - | 4 | 410 | |||
AY550930.2 | - | 3 | 327 | |||
BC029702.1 | - | 4 | 408 | 5053250 |