Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpm6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177955.1 | NP_001171426.1 | 4 | 921 | ||
NM_001177956.1 | NP_001171427.1 | 4 | 921 | |||
NM_001177957.1 | NP_001171428.1 | 4 | 921 | |||
NM_001177958.1 | NP_001171429.1 | 3 | 801 | |||
NM_001177959.1 | NP_001171430.1 | 3 | 801 | |||
NM_001177960.1 | NP_001171431.1 | 3 | 801 | |||
NM_001177961.1 | NP_001171432.1 | 3 | 466 | |||
NM_001177962.1 | NP_001171433.1 | 3 | 466 | |||
NM_023122.3 | NP_075611.1 | 3 | 466 | |||
GenBank mRNA | AF254869.1 | - | 3 | 662 | ||
AF254870.1 | - | 4 | 782 | |||
AF254871.1 | - | 3 | 437 | |||
AF254872.1 | - | 3 | 662 | |||
AF254873.1 | - | 3 | 662 | |||
AF254874.1 | - | 4 | 782 | |||
AF254875.1 | - | 4 | 782 | |||
AF254876.1 | - | 3 | 437 | |||
AF254877.1 | - | 3 | 437 | |||
AF254878.1 | - | 4 | 809 | |||
AF254879.1 | - | 5 | 929 | |||
AK016567.1 | - | 3 | 391 | |||
AK132778.1 | - | 3 | 453 | |||
AK140714.1 | - | 3 | 481 | |||
BC003912.1 | - | 2 | 249 | |||
BC025165.1 | - | 3 | 427 | 3981536 | ||
S65737.1 | - | 3 | 458 |