Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Palm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161747.1 | NP_001155219.1 | 3 | 247 | ||
NM_023128.4 | NP_075617.3 | 3 | 247 | |||
GenBank mRNA | AK009200.1 | - | 2 | 88 | ||
AK037044.1 | - | 3 | 104 | |||
AK082163.1 | - | 3 | 249 | |||
AK133733.1 | - | 3 | 153 | |||
AK162411.1 | - | 3 | 240 | |||
BC015297.1 | - | 3 | 231 | 4223845 | ||
Y14771.1 | - | 3 | 210 |