Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rce1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023131.1 | NP_075620.1 | 3 | 300 | ||
GenBank mRNA | AJ251645.1 | - | 3 | 300 | ||
AK012946.1 | - | 3 | 256 | |||
BC004060.1 | - | 1, 2 | 149 | 3590170 | ||
BC048686.1 | - | 3 | 324 |