Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tor3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023141.2 | NP_075630.2 | 2 | 359 | ||
GenBank mRNA | AJ299404.1 | - | 2 | 327 | ||
AK007814.1 | - | 2 | 348 | |||
AK009693.1 | - | 2 | 377 | |||
AK017908.1 | - | 2 | 304 | |||
AK151048.1 | - | 2 | 345 | |||
AK151390.1 | - | 2 | 326 | |||
AK152243.1 | - | 2 | 324 | |||
AK152358.1 | - | 2 | 360 | |||
AK152484.1 | - | 2 | 360 | |||
AK153391.1 | - | 2 | 345 | |||
AK170296.1 | - | 2 | 354 | |||
AK172355.1 | - | 2 | 334 | |||
BC025525.1 | - | 2 | 344 | |||
BC052851.1 | - | 2 | 357 | |||
BC071216.1 | - | 2 | 340 | 30439199 |