Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ranbp17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023146.2 | NP_075635.1 | 2 | 222 | ||
GenBank mRNA | AJ271458.1 | - | 2 | 120 | ||
AK016583.1 | - | 2 | 122 | |||
AK035897.1 | - | 2 | 119 | |||
AK046865.1 | - | 2 | 107 | |||
AK139704.1 | - | 2 | 131 | |||
AK163271.1 | - | 121 | ||||
BC033503.1 | - | 2 | 116 | |||
BY731762.1 | - | 2 | 223 |