Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cndp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289531.1 | NP_001276460.1 | 2 | 285 | ||
NM_023149.3 | NP_075638.2 | 3 | 253 | |||
GenBank mRNA | AB046738.1 | - | 3 | 194 | ||
AK003779.1 | - | 3 | 174 | |||
AK131124.1 | - | 2 | 165 | |||
AK149751.1 | - | 2 | 209 | |||
AK151013.1 | - | 3 | 175 | |||
AK152442.1 | - | 2 | 251 | |||
AK152729.1 | - | 2 | 223 | |||
AK153040.1 | - | 2 | 223 | |||
AK159129.1 | - | 2 | 251 | |||
AK159739.1 | - | 3 | 181 | |||
AK168029.1 | - | 3 | 192 | |||
AK168652.1 | - | 3 | 204 | |||
AK172034.1 | - | 3 | 175 | |||
AK191043.1 | - | 1 | 20 | |||
AK200700.1 | - | 1 | 70 | |||
BC005532.1 | - | 3 | 184 | 3496319 | ||
CJ078789.1 | - | 3 | 253 | |||
CN721222.1 | - | 2 | 285 |