Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085472.2 | NP_001078941.2 | 4 | 263 | ||
NM_001085473.2 | NP_001078942.2 | 10 | 2624 | |||
NM_001242605.1 | NP_001229534.1 | 3 | 327 | |||
NM_001242606.1 | NP_001229535.1 | 3 | 327 | |||
NM_019567.3 | NP_062513.3 | 4 | 392 | |||
NM_023190.3 | NP_075679.2 | 10 | 2624 | |||
GenBank mRNA | AF124725.1 | - | 4 | 323 | ||
AF124729.1 | - | 3 | 258 | |||
AF168782.1 | - | 10 | 2625 | |||
AK012099.1 | - | 4 | 259 | |||
AK030449.1 | - | 10 | 2317 | |||
AK122342.1 | - | 10 | 2295 | |||
AV510393.1 | - | 4 | 259 | |||
BC019127.1 | - | 3 | 312 | |||
BC052755.1 | - | 3 | 248 | |||
BC075691.1 | - | 10 | 2309 | |||
BC094217.1 | - | 3 | 309 | |||
BY287113.1 | - | 3 | 199 | |||
BY711288.1 | - | 4 | 259 |