Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pbk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023209.2 | NP_075698.1 | 4 | 412 | ||
GenBank mRNA | AB041882.1 | - | 4 | 415 | ||
AK003838.1 | - | 4 | 373 | |||
AK076121.1 | - | 4 | 485 | |||
BC020099.1 | - | 4 | 418 | 3673060 |