Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023214.7 | NP_075703.1 | 4, 5 | 576 | ||
GenBank mRNA | AF233322.1 | - | 4, 5 | 438 | ||
AF529196.1 | - | 4, 5 | 384 | |||
AK010008.1 | - | 4, 5 | 576 | |||
AK075802.1 | - | 4, 5 | 549 | |||
BC017136.1 | - | 4, 5 | 526 | 4911068 | ||
BC050193.1 | - | 4, 5 | 475 | 30019802 |