Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022981.4 | NP_075357.4 | 6 | 1164 | ||
GenBank mRNA | AJ242914.1 | - | 6 | 919 | ||
AK013583.1 | - | 6 | 970 | |||
AK040989.1 | - | 6 | 898 | |||
AK202349.1 | - | 2 | 188 | |||
BC006657.1 | - | 6 | 913 | 3500984 |