Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl6ip5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022992.2 | NP_075368.1 | 3 | 553 | ||
GenBank mRNA | AF265214.1 | - | 3 | 524 | ||
AF421860.1 | - | 3 | 559 | |||
AK005259.1 | - | 3 | 550 | |||
AK008519.1 | - | 3 | 533 | |||
AK076519.1 | - | 3 | 550 | |||
AK077826.1 | - | 3 | 559 | |||
BC003897.1 | - | 3 | 553 | 3600236 | ||
D87211.1 | - | 3 | 596 |