Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pmepa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022995.3 | NP_075371.2 | 4 | 1155 | ||
GenBank mRNA | AF220208.1 | - | 3 | 590 | ||
AK008976.1 | - | 4 | 1154 | |||
AK136450.1 | - | 4 | 1156 | |||
BC023092.1 | - | 4 | 828 | |||
BC036995.1 | - | 4 | 851 | |||
BC069890.1 | - | 806 | ||||
BC092094.1 | - | 4 | 810 |