Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCN8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177984.2 | NP_001171455.1 | 13 | 2268 | ||
NM_014191.3 | NP_055006.1 | 13 | 2268 | |||
XM_006719556.3 | XP_006719619.1 | 13 | 2859 | |||
XM_011538650.2 | XP_011536952.1 | 13 | 2287 | |||
XM_011538651.2 | XP_011536953.1 | 13 | 2252 | |||
XM_017019794.1 | XP_016875283.1 | 13 | 2265 | |||
XM_017019795.1 | XP_016875284.1 | 13 | 2284 | |||
XM_017019796.1 | XP_016875285.1 | 13 | 2284 | |||
GenBank mRNA | AB027567.1 | - | 13 | 2268 | ||
AF225988.1 | - | 12 | 2090 | |||
AK309213.1 | - | 13 | 2218 | |||
FJ611941.1 | - | 12 | 2090 |