Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rce1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023131.1 | NP_075620.1 | 8 | 1160 | ||
GenBank mRNA | AK012946.1 | - | 8 | 1116 | ||
BC004060.1 | - | 7 | 1009 | 3590170 | ||
BC048686.1 | - | 8 | 1209 |