Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nono | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252518.1 | NP_001239447.1 | 4 | 388 | ||
NM_023144.2 | NP_075633.2 | 4 | 391 | |||
GenBank mRNA | AK004830.1 | - | 4 | 354 | ||
AK013444.1 | - | 4 | 305 | |||
AK028338.1 | - | 4 | 356 | |||
AK033201.1 | - | 4 | 317 | |||
AK145179.1 | - | 4 | 364 | |||
AK161152.1 | - | 4 | 375 | |||
AK165855.1 | - | 4 | 350 | |||
AK167297.1 | - | 4 | 322 | |||
AK169202.1 | - | 4 | 354 | |||
AK169303.1 | - | 4 | 375 | |||
AK213551.1 | - | 2 | 214 | |||
BC005465.1 | - | 4 | 350 | 3600935 | ||
BC083074.1 | - | 2 | 228 | 6390386 | ||
CJ180326.1 | - | 4 | 391 | |||
CT010241.1 | - | 2 | 210 | |||
S64860.1 | - | 4 | 338 |