Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cndp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289531.1 | NP_001276460.1 | 3, 4 | 537 | ||
NM_023149.3 | NP_075638.2 | 4, 5 | 505 | |||
GenBank mRNA | AB046738.1 | - | 4, 5 | 446 | ||
AK003779.1 | - | 4, 5 | 426 | |||
AK131124.1 | - | 3, 4 | 417 | |||
AK149751.1 | - | 3, 4 | 461 | |||
AK151013.1 | - | 4, 5 | 427 | |||
AK152442.1 | - | 3, 4 | 503 | |||
AK152729.1 | - | 3, 4 | 475 | |||
AK153040.1 | - | 3, 4 | 476 | |||
AK159129.1 | - | 3, 4 | 503 | |||
AK159739.1 | - | 4, 5 | 433 | |||
AK168029.1 | - | 4, 5 | 444 | |||
AK168652.1 | - | 4, 5 | 456 | |||
AK168856.1 | - | 2, 3 | 194 | |||
AK172034.1 | - | 4, 5 | 427 | |||
AK189897.1 | - | 1, 2 | 73 | |||
BC005532.1 | - | 4, 5 | 436 | 3496319 | ||
CN721222.1 | - | 3, 4 | 537 |