Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023167.2 | NP_075656.2 | 6, 7 | 764 | ||
GenBank mRNA | AB041599.1 | - | 5, 6 | 516 | ||
AB049634.1 | - | 6, 7 | 763 | |||
AK002464.1 | - | 6, 7 | 600 | |||
AK003746.2 | - | 6, 7 | 721 | |||
BC023078.1 | - | 5 | 1124 | |||
BC037064.1 | - | 6, 7 | 597 | |||
BC039983.1 | - | 6, 7 | 732 | 4952757 | ||
BC051685.1 | - | 6, 7 | 577 | |||
BC061095.1 | - | 6, 7 | 580 | 6588836 |