Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085472.2 | NP_001078941.2 | 6 | 450 | ||
NM_001242605.1 | NP_001229534.1 | 5 | 514 | |||
NM_019567.3 | NP_062513.3 | 6 | 579 | |||
NM_023190.3 | NP_075679.2 | 12 | 2811 | |||
GenBank mRNA | AF124725.1 | - | 6 | 510 | ||
AF124729.1 | - | 5 | 445 | |||
AF168782.1 | - | 12 | 2812 | |||
AK012099.1 | - | 6 | 443 | |||
AK030449.1 | - | 12 | 2505 | |||
AV510393.1 | - | 6 | 446 | |||
BC019127.1 | - | 5 | 499 | |||
BC052755.1 | - | 5 | 435 | |||
BY287113.1 | - | 5 | 383 | |||
BY711288.1 | - | 6 | 443 |