Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nif3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022988.2 | NP_075364.2 | 6 | 1026 | ||
GenBank mRNA | AF284439.1 | - | 7 | 1503 | ||
AK003978.1 | - | 7 | 1134 | |||
AK011670.1 | - | 7 | 1043 | |||
AK131631.1 | - | 7 | 1389 | |||
AK154815.1 | - | 7 | 1118 | |||
AK158616.1 | - | 7 | 1409 | |||
BC065163.1 | - | 6 | 1026 | 6404016 | ||
BC089475.1 | - | 7 | 953 | 30504614 |