Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpo7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023045.2 | NP_075532.1 | 2 | 194 | ||
GenBank mRNA | AJ297360.1 | - | 2 | 100 | ||
AK035775.1 | - | 2 | 119 | |||
AK041768.1 | - | 2 | 102 | |||
AK051105.1 | - | 2 | 116 | |||
AK122361.1 | - | 1 | 79 | |||
AK143729.1 | - | 2 | 163 | |||
AK164595.1 | - | 2 | 122 | |||
BC029702.1 | - | 2 | 120 | 5053250 | ||
BY034562.1 | - | 2 | 195 |