Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252533.1 | NP_001239462.1 | 13 | 2776 | ||
NM_023116.4 | NP_075605.1 | 14 | 2907 | |||
GenBank mRNA | AK034054.1 | - | 14 | 2908 | ||
BC048944.1 | - | 1 | 54 | |||
BC056183.1 | - | 1 | 164 | |||
CX565964.1 | - | 1 | 704 |