Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc25b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111075.4 | NP_001104545.1 | 7 | 945 | ||
NM_023117.4 | NP_075606.1 | 8 | 1023 | |||
GenBank mRNA | AK004836.1 | - | 7 | 902 | ||
AK043730.1 | - | 7 | 936 | |||
AK153903.1 | - | 8 | 1024 | |||
AK157397.1 | - | 7 | 903 | |||
BC048164.1 | - | 7 | 942 | |||
BC057568.1 | - | 7 | 816 | 6401489 | ||
S93521.1 | - | 8 | 892 |