Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnb1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081682.2 | NP_001075151.1 | 7, 8 | 987 | ||
NM_001285491.1 | NP_001272420.1 | 7, 8 | 983 | |||
NM_001285493.1 | NP_001272422.1 | 8, 9 | 1022 | |||
NM_001285494.1 | NP_001272423.1 | 7, 8 | 887 | |||
NM_023120.5 | NP_075609.2 | 7, 8 | 929 | |||
GenBank mRNA | AF301595.1 | - | 7, 8 | 880 | ||
AK010489.1 | - | 7, 8 | 887 | |||
AK016952.1 | - | 7, 8 | 913 | |||
AK087954.1 | - | 7, 8 | 982 | |||
AK158210.1 | - | 8, 9 | 1009 | |||
AK166195.1 | - | 7, 8 | 871 | |||
AK171253.1 | - | 7, 8 | 909 | |||
AK173227.1 | - | 7, 8 | 912 | |||
BC024635.1 | - | 8, 9 | 1023 | 4193427 | ||
BC037676.1 | - | 6, 7 | 815 | 3493788 |
Set Membership: |
G-protein coupled receptor |