Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Renbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164704.1 | NP_001158176.1 | 11 | 1355 | ||
NM_023132.3 | NP_075621.3 | 11 | 1397 | |||
GenBank mRNA | AK014347.1 | - | 11 | 1317 | ||
AK146652.1 | - | 11 | 1362 | |||
AK167648.1 | - | 11 | 1268 | |||
AY183138.1 | - | 11 | 1365 | |||
BC014821.1 | - | 11 | 1291 | 4238244 | ||
BC034846.1 | - | 7 | 2081 | |||
BC035455.1 | - | 11 | 1291 |