Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cndp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289531.1 | NP_001276460.1 | 11 | 1955 | ||
NM_023149.3 | NP_075638.2 | 12 | 1923 | |||
GenBank mRNA | AB046738.1 | - | 12 | 1864 | ||
AK003779.1 | - | 12 | 1844 | |||
AK131124.1 | - | 11 | 1834 | |||
AK149751.1 | - | 11 | 1879 | |||
AK151013.1 | - | 12 | 1845 | |||
AK152442.1 | - | 11 | 1920 | |||
AK152729.1 | - | 11 | 1893 | |||
AK153040.1 | - | 11 | 1894 | |||
AK159129.1 | - | 11 | 1922 | |||
AK159739.1 | - | 12 | 1850 | |||
AK168029.1 | - | 12 | 1862 | |||
AK168652.1 | - | 12 | 1874 | |||
AK168856.1 | - | 10 | 1612 | |||
AK194231.1 | - | 1 | 174 | |||
BC005532.1 | - | 12 | 1854 | 3496319 | ||
CJ306472.1 | - | 1 | 215 |