Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cwc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023153.3 | NP_075642.1 | 5, 6 | 620 | ||
GenBank mRNA | AJ250394.1 | - | 5, 6 | 511 | ||
AK002864.1 | - | 5, 6 | 559 | |||
AK003202.1 | - | 5, 6 | 552 | |||
AK003591.1 | - | 3, 4 | 262 | |||
AK003834.2 | - | 5, 6 | 565 | |||
AK145249.1 | - | 5, 6 | 560 | |||
AK167776.1 | - | 5, 6 | 540 | |||
AK168365.1 | - | 5, 6 | 540 | |||
AY842452.1 | - | 4, 5 | 466 | |||
BC004726.1 | - | 5, 6 | 556 | 3582653 |