Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023167.2 | NP_075656.2 | 8 | 918 | ||
GenBank mRNA | AB041599.1 | - | 7 | 670 | ||
AB049634.1 | - | 8 | 917 | |||
AK002464.1 | - | 8 | 754 | |||
AK003746.2 | - | 8 | 875 | |||
BC023078.1 | - | 5 | 1360 | |||
BC037064.1 | - | 8 | 751 | |||
BC039983.1 | - | 8 | 886 | 4952757 | ||
BC051685.1 | - | 8 | 731 | |||
BC061095.1 | - | 8 | 734 | 6588836 |