Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pbk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023209.2 | NP_075698.1 | 7 | 930 | ||
GenBank mRNA | AB041882.1 | - | 7 | 933 | ||
AK003838.1 | - | 7 | 891 | |||
AK076121.1 | - | 7 | 1003 | |||
BC006754.1 | - | 3 | 295 | |||
BC020099.1 | - | 7 | 936 | 3673060 |