Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgpep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023217.4 | NP_075706.1 | 4, 5 | 540 | ||
GenBank mRNA | AJ278829.1 | - | 4, 5 | 472 | ||
AK003373.1 | - | 4, 5 | 484 | |||
AK012658.1 | - | 4, 5 | 472 | |||
AK047835.1 | - | 4, 5 | 484 | |||
AK049632.1 | - | 4, 5 | 463 | |||
BC043041.1 | - | 4, 5 | 450 | |||
BC051938.1 | - | 4, 5 | 469 | 5717365 |