Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sppl2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023220.2 | NP_075709.2 | 12, 13 | 1492 | ||
GenBank mRNA | AB041547.1 | - | 12, 13 | 1492 | ||
AK155011.1 | - | 12, 13 | 1291 |