Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Palmd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023245.3 | NP_075734.3 | 2 | 514 | ||
GenBank mRNA | AF263246.1 | - | 2 | 218 | ||
AJ312215.1 | - | 2 | 237 | |||
AK014531.1 | - | 2 | 312 | |||
AK136939.1 | - | 2 | 421 | |||
AK171869.1 | - | 2 | 245 | |||
AL391037.1 | - | 2 | 218 | |||
BC010193.1 | - | 2 | 278 | 3156945 |