Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ypel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291047.1 | NP_001277976.1 | 5 | 711 | ||
NM_001291053.1 | NP_001277982.1 | 3 | 305 | |||
NM_023249.6 | NP_075738.1 | 4 | 585 | |||
GenBank mRNA | AB098740.1 | - | 3 | 197 | ||
AF190624.1 | - | 4 | 501 | |||
AK005955.1 | - | 4 | 421 | |||
AK006049.1 | - | 4 | 421 | |||
AK006137.1 | - | 3 | 203 | |||
AK006505.1 | - | 4 | 396 | |||
AK006858.1 | - | 4 | 473 | |||
AK011452.1 | - | 4 | 516 | |||
AK014931.1 | - | 4 | 411 | |||
AK015752.1 | - | 3 | 422 | |||
AK018910.1 | - | 4 | 396 | |||
AK019819.1 | - | 4 | 416 | |||
AK076063.1 | - | 4 | 528 | |||
AK133401.1 | - | 4 | 499 | |||
AK139453.1 | - | 4 | 493 | |||
BB649357.1 | - | 4 | 585 | |||
BC049737.1 | - | 3 | 215 | 6771362 | ||
BQ573264.1 | - | 3 | 271 |