Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qsox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024945.1 | NP_001020116.1 | 1 | 360 | ||
NM_023268.2 | NP_075757.1 | 1 | 360 | |||
GenBank mRNA | AB044284.1 | - | 1 | 268 | ||
AK004880.1 | - | 1 | 306 | |||
AK083938.1 | - | 1 | 311 | |||
AK149465.1 | - | 1 | 295 | |||
AK163119.1 | - | 1 | 298 | |||
AK166839.1 | - | 1 | 306 | |||
AK169877.1 | - | 1 | 313 | |||
AK169920.1 | - | 1 | 309 | |||
AK170449.1 | - | 1 | 273 | |||
AK171127.1 | - | 1 | 284 | |||
BC076590.1 | - | 1 | 314 | 30548238 |