Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nuf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023284.3 | NP_075773.2 | 2 | 239 | ||
GenBank mRNA | AF326732.1 | - | 3 | 280 | ||
AK010351.1 | - | 2 | 161 | |||
AK012011.1 | - | 2 | 173 | |||
AK044882.1 | - | 2 | 3558 | |||
AK089939.1 | - | 3 | 266 | |||
AK132467.1 | - | 2 | 240 | |||
AK162249.1 | - | 3 | 275 | |||
BC020026.1 | - | 3 | 248 | 3587655 |