Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nde1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114085.1 | NP_001107557.1 | 5 | 620 | ||
NM_001285503.1 | NP_001272432.1 | 5 | 620 | |||
NM_001285504.1 | NP_001272433.1 | 4 | 471 | |||
NM_023317.2 | NP_075806.2 | 5 | 620 | |||
GenBank mRNA | AF290473.1 | - | 3 | 354 | ||
AF322073.1 | - | 4 | 527 | |||
AK012830.1 | - | 5 | 550 | |||
AK012861.1 | - | 5 | 570 | |||
AK037289.1 | - | 5 | 563 | |||
AK146982.1 | - | 5 | 550 | |||
AK150826.1 | - | 5 | 610 | |||
AK166858.1 | - | 4 | 450 | |||
AK169743.1 | - | 5 | 550 | |||
BC023267.1 | - | 5 | 599 | 3672565 |