Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023371.3 | NP_075860.1 | 2 | 249 | ||
GenBank mRNA | AB009691.1 | - | 2 | 268 | ||
AK002665.1 | - | 2 | 275 | |||
AK003369.1 | - | 2 | 250 | |||
AK003931.1 | - | 2 | 263 | |||
AK054045.1 | - | 2 | 250 | |||
AK139395.1 | - | 2 | 248 | |||
AK150652.1 | - | 2 | 250 | |||
AK160228.1 | - | 2 | 262 | |||
AK206050.1 | - | 1 | 14 | |||
BC038254.1 | - | 2 | 256 | 4971446 |
Gene Symbol: Pin1rt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033768.2 | NP_001028940.1 | 2 | 444 | ||
GenBank mRNA | AK145372.1 | - | 2 | 445 | ||
BC147362.1 | - | 2 | 266 | 9056420 | ||
BC147363.1 | - | 2 | 266 | 9056439 |