Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023418.2 | NP_075907.2 | 2 | 474 | ||
GenBank mRNA | AF283667.1 | - | 2 | 386 | ||
AK004921.1 | - | 2 | 330 | |||
AK009905.2 | - | 2 | 327 | |||
AK144681.1 | - | 2 | 328 | |||
AK150716.1 | - | 2 | 323 | |||
AK152443.1 | - | 2 | 323 | |||
AK153202.1 | - | 2 | 327 | |||
AK159305.1 | - | 2 | 327 | |||
AK165410.1 | - | 2 | 323 | |||
AK168949.1 | - | 2 | 320 | |||
AK180741.1 | - | 1 | 71 | |||
AK192398.1 | - | 1 | 105 | |||
AK211756.1 | - | 1 | 19 | |||
AK212069.1 | - | 1 | 19 | |||
AK214477.1 | - | 1 | 19 | |||
AK216443.1 | - | 1 | 19 | |||
BC002241.1 | - | 2 | 422 | 3491964 | ||
BC005661.1 | - | 2 | 388 | 3588761 | ||
BC066844.1 | - | 2 | 311 | 30471755 | ||
BC067412.1 | - | 1 | 127 | |||
BC083090.1 | - | 2 | 323 | 30444229 | ||
BC106139.1 | - | 2 | 394 | 6528521 |