Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023479.2 | NP_075968.2 | 2, 3 | 348 | ||
GenBank mRNA | AF308696.2 | - | 2, 3 | 286 | ||
AK156730.1 | - | 2, 3 | 349 | |||
AK172604.1 | - | 1, 2 | 717 | |||
AK211653.1 | - | 1, 2 | 44 | |||
BC023134.1 | - | 2, 3 | 292 | |||
BC048235.1 | - | 2, 3 | 282 | 6493124 |