Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023514.4 | NP_076003.3 | 7 | 655 | ||
GenBank mRNA | AJ250193.1 | - | 3 | 124 | ||
AK009082.1 | - | 7 | 605 | |||
AK049465.1 | - | 7 | 613 | |||
AK144656.1 | - | 7 | 613 | |||
AK150985.1 | - | 7 | 612 | |||
AK185376.1 | - | 2 | 106 | |||
BC022587.1 | - | 2 | 93 | |||
BC051936.1 | - | 6 | 348 | |||
BC069980.1 | - | 5 | 249 | |||
BC083159.1 | - | 6 | 416 | |||
BC116437.2 | - | 7 | 605 | 40104493 |