Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASIC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182847.2 | NP_878267.2 | 9 | 2100 | ||
XM_005246671.2 | XP_005246728.2 | 10 | 2014 | |||
XM_017004439.1 | XP_016859928.1 | 10 | 1173 | |||
GenBank mRNA | BC010439.1 | - | 9 | 1940 | 4215164 |