Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023223.2 | NP_075712.2 | 2, 3 | 560 | ||
GenBank mRNA | AB045313.1 | - | 2, 3 | 464 | ||
AF312208.1 | - | 2, 3 | 457 | |||
AK011723.1 | - | 2, 3 | 559 | |||
AK029424.1 | - | 2, 3 | 512 | |||
AK075998.1 | - | 2, 3 | 508 | |||
AK076030.1 | - | 2, 3 | 512 | |||
AK083459.1 | - | 2, 3 | 389 | |||
AK160204.1 | - | 2, 3 | 572 | |||
AK168650.1 | - | 2, 3 | 512 | |||
BC003215.1 | - | 2, 3 | 497 | 3157842 | ||
BY736811.1 | - | 2, 3 | 567 |